CAS RN 131738-73-3
IUPAC Name:
1-bromo-3-(propan-2-yloxy)benzene
Synonyms:
1-bromo-3-isopropoxybenzene
Molecular Weight (g/mol):
215.09
Molecular Formula:
C9H11BrO
InChi Key:
JYWJZIQSPRFCDB-UHFFFAOYSA-N
SMILES:
CC(C)OC1=CC(Br)=CC=C1
1
–
1
of
1
results
1
1-Bromo-3-isopropoxybenzene 98.0+%, TCI America™
CAS: 131738-73-3 Molecular Formula: C9H11BrO Molecular Weight (g/mol): 215.09 MDL Number: MFCD00070756 InChI Key: JYWJZIQSPRFCDB-UHFFFAOYSA-N Synonym: 1-bromo-3-isopropoxybenzene,3-bromophenyl isopropyl ether,1-bromo-3-propan-2-yloxy benzene,3-bromo-isopropoxybenzene,3-bromoisoproproxylbenzene,2-3-bromophenoxy propane,3-bromoisopropoxybenzene,3-bromo phenyl isopropyl ether,benzene, 1-bromo-3-1-methylethoxy,pubchem14867 PubChem CID: 3260801 IUPAC Name: 1-bromo-3-(propan-2-yloxy)benzene SMILES: CC(C)OC1=CC(Br)=CC=C1