missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene 98.0+%, TCI America™

Catalog No. p-7123179
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38081G 1 g
B38085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38081G Supplier TCI America Supplier No. B38081G

Chemical Identifiers

CAS 105529-58-6
Molecular Formula C7H3BrF4O
Molecular Weight (g/mol) 258.998
MDL Number MFCD01861132
InChI Key SBSFDYRKNUCGBZ-UHFFFAOYSA-N
Synonym 4-bromo-2-fluoro-1-trifluoromethoxy benzene, 1-bromo-3-fluoro-4-trifluoromethoxy benzene, 1-bromo-3-fluoro-4-trifluoromethoxybenzene, 2-fluoro-4-bromotrifluoromethoxybenzene, 4-bromo-2-fluorotrifluoromethoxybenzene, benzene, 4-bromo-2-fluoro-1-trifluoromethoxy, 3-fluoro-4-trifluoromethoxy bromobenzene, 3-bromo-2-fluorotrifluoromethoxybenzene, 4-bromo-2-fluorophenyl trifluoromethyl ether, 4-bromo-alpha,alpha,alpha,2-tetrafluoroanisole
PubChem CID 2782217
IUPAC Name 4-bromo-2-fluoro-1-(trifluoromethoxy)benzene
SMILES C1=CC(=C(C=C1Br)F)OC(F)(F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 259.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene