accessibility menu, dialog, popup

UserName

CAS RN 105529-58-6

IUPAC Name: 4-bromo-2-fluoro-1-(trifluoromethoxy)benzene
Synonyms: 4-bromo-2-fluoro-1-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 259
Molecular Formula: C7H3BrF4O
InChi Key: SBSFDYRKNUCGBZ-UHFFFAOYSA-N
SMILES: FC1=C(OC(F)(F)F)C=CC(Br)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results