missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-fluoro-4-iodobenzene, 99+%

Catalog No. AC249400250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC249400250 25 g Glass bottle
AC249401000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC249400250 Supplier Thermo Scientific Chemicals Supplier No. 249400250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 105931-73-5 | C6H3BrFI | 300.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105931-73-5
Molecular Formula C6H3BrFI
Molecular Weight (g/mol) 300.90
MDL Number MFCD00010608
InChI Key XRMZKCQCINEBEI-UHFFFAOYSA-N
Synonym 1-bromo-3-fluoro-4-iodobenzene, 3-fluoro-4-iodobromobenzene, 2-fluoro-4-bromoiodobenzene, 4-bromo-2-fluoroiodobenzene, 4-bromo-2-fluoro-1-iodo-benzene, benzene, 4-bromo-2-fluoro-1-iodo, pubchem3232, acmc-2098hz, 2-fluoro-4-bromo iodobenzene, 3-fluoro-4-iodo-bromobenzene
PubChem CID 2724516
IUPAC Name 4-bromo-2-fluoro-1-iodobenzene
SMILES FC1=C(I)C=CC(Br)=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 46°C to 49°C
Color White to Beige
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(F)I
Quantity 25 g
Formula Weight 300.89
Percent Purity 99+%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1-Bromo-3-fluoro-4-iodobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only