missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-fluoro-2-nitrobenzene 98.0+%, TCI America™

Catalog No. B48425G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48425G 5 g
B48421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48425G Supplier TCI America Supplier No. B48425G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 886762-70-5
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 220.00
MDL Number MFCD07368788
InChI Key VFPAOFBPEYCAAZ-UHFFFAOYSA-N
PubChem CID 2783152
IUPAC Name 1-bromo-3-fluoro-2-nitrobenzene
SMILES [O-][N+](=O)C1=C(F)C=CC=C1Br

Specifications

Melting Point 40°C
Color Yellow
Quantity 5 g
Formula Weight 220.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Bromo-3-fluoro-2-nitrobenzene
TSCA No
Recommended Storage Refrigerator