accessibility menu, dialog, popup

UserName

CAS RN 886762-70-5

BrNOOF

CAS RN 886762-70-5

IUPAC Name: 1-bromo-3-fluoro-2-nitrobenzene
Synonyms: 1-bromo-3-fluoro-2-nitrobenzene
Molecular Weight (g/mol): 220
Molecular Formula: C6H3BrFNO2
InChi Key: VFPAOFBPEYCAAZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(F)C=CC=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results