missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chloro-2-iodobenzene, 97%

Catalog No. p-7056862
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66020MD 250 mg
AAH6602003 1 g
AAH6602006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66020MD Supplier Thermo Scientific Chemicals Supplier No. H66020MD

CAS: 450412-28-9 | C6H3BrClI | 317.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 450412-28-9
Molecular Formula C6H3BrClI
Molecular Weight (g/mol) 317.35
MDL Number MFCD09907935
InChI Key GIMVCZMZRZGDTL-UHFFFAOYSA-N
Synonym 2-bromo-6-chloroiodobenzene, 1-bromo-2-iodo-3-chlorobenzene, benzene, 1-?bromo-?3-?chloro-?2-?iodo
PubChem CID 10335963
IUPAC Name 1-bromo-3-chloro-2-iodobenzene
SMILES ClC1=C(I)C(Br)=CC=C1

Specifications

Quantity 250 mg
Beilstein 9119447
Sensitivity Light sensitive
Formula Weight 317.35
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Bromo-3-chloro-2-iodobenzene
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only