CAS RN 450412-28-9
IUPAC Name:
1-bromo-3-chloro-2-iodobenzene
Synonyms:
1-bromo-3-chloro-2-iodobenzene
Molecular Weight (g/mol):
317.35
Molecular Formula:
C6H3BrClI
InChi Key:
GIMVCZMZRZGDTL-UHFFFAOYSA-N
SMILES:
ClC1=C(I)C(Br)=CC=C1
1
–
1
of
1
results
1
Promotions Available
1-Bromo-3-chloro-2-iodobenzene, 97%
1-Bromo-3-chloro-2-iodobenzene, 97%
CAS: 450412-28-9 Molecular Formula: C6H3BrClI Molecular Weight (g/mol): 317.35 MDL Number: MFCD09907935 InChI Key: GIMVCZMZRZGDTL-UHFFFAOYSA-N Synonym: 2-bromo-6-chloroiodobenzene,1-bromo-2-iodo-3-chlorobenzene,benzene, 1-?bromo-?3-?chloro-?2-?iodo PubChem CID: 10335963 IUPAC Name: 1-bromo-3-chloro-2-iodobenzene SMILES: ClC1=C(I)C(Br)=CC=C1