missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chloro-2-fluorobenzene, 95%

Catalog No. AAH6162214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6162214 25 g
AAH6162222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6162214 Supplier Thermo Scientific Chemicals Supplier No. H6162214

CAS: 144584-65-6 | C6H3BrClF | 209.442 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144584-65-6
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD03412182
InChI Key DRNWNTAANHEQMK-UHFFFAOYSA-N
Synonym 3-chloro-2-fluorobromobenzene, 1-bromo-2-fluoro-3-chlorobenzene, 1-bromo-3-chloro-2-fluoro-benzene, pubchem4338, acmc-1c0jw, intermediates-zcf02109, ksc493i6p, benzene,1-bromo-3-chloro-2-fluoro
PubChem CID 3685762
IUPAC Name 1-bromo-3-chloro-2-fluorobenzene
SMILES C1=CC(=C(C(=C1)Br)F)Cl

Specifications

Melting Point 42°C to 46°C
Color White to Yellow
Quantity 25 g
Formula Weight 209.44
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 1-Bromo-3-chloro-2-fluorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only