accessibility menu, dialog, popup

UserName

BrFCl

CAS RN 144584-65-6

IUPAC Name: 1-bromo-3-chloro-2-fluorobenzene
Synonyms: 1-bromo-3-chloro-2-fluorobenzene
Molecular Weight (g/mol): 209.44
Molecular Formula: C6H3BrClF
InChi Key: DRNWNTAANHEQMK-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=CC=C1Br
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)