missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-dimethoxybenzene 98.0+%, TCI America™

Catalog No. p-7123201
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38485G 5 g
B384825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38485G Supplier TCI America Supplier No. B38485G
Only null left

Chemical Identifiers

CAS 20469-65-2
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD06200685
InChI Key KRWRFIMBWRVMKE-UHFFFAOYSA-N
Synonym 3,5-dimethoxybromobenzene, benzene, 1-bromo-3,5-dimethoxy, 1,3-dimethoxy-5-bromobenzene, 1-bromo-3,5-dimethoxy-benzene, 1-bromo-3,5-dimethox, 3,5-dimethoxy-bromobenzene, bromo-3,5-dimethoxybenzene, 3,5-dimethoxy bromobenzene, acmc-1cs59, ksc495e7l
PubChem CID 639187
IUPAC Name 1-bromo-3,5-dimethoxybenzene
SMILES COC1=CC(OC)=CC(Br)=C1

Specifications

Melting Point 67°C
Color White-Yellow
Quantity 5 g
Formula Weight 217.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Bromo-3,5-dimethoxybenzene