accessibility menu, dialog, popup

UserName

H3COBrOCH3

CAS RN 20469-65-2

IUPAC Name: 1-bromo-3,5-dimethoxybenzene
Synonyms: 1-bromo-3,5-dimethoxybenzene
Molecular Weight (g/mol): 217.06
Molecular Formula: C8H9BrO2
InChi Key: KRWRFIMBWRVMKE-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(Br)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)