missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-dimethoxybenzene, 97%

Catalog No. p-7036864
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5531206 5 g
AAH5531214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5531206 Supplier Thermo Scientific Chemicals Supplier No. H5531206
Only null left

CAS: 20469-65-2 | C8H9BrO2 | 217.06 g/mol

1-Bromo-3,5-dimethoxybenzene is used as an intermediate in the synthetic preparation of pharmaceutical inhibitors via cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3,5-dimethoxybenzene is used as an intermediate in the synthetic preparation of pharmaceutical inhibitors via cross-coupling reactions.

Solubility
Soluble in methanol.

Notes
Reacts with strong oxidizing agents.

Chemical Identifiers

CAS 20469-65-2
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD06200685
InChI Key KRWRFIMBWRVMKE-UHFFFAOYSA-N
Synonym 3,5-dimethoxybromobenzene, benzene, 1-bromo-3,5-dimethoxy, 1,3-dimethoxy-5-bromobenzene, 1-bromo-3,5-dimethoxy-benzene, 1-bromo-3,5-dimethox, 3,5-dimethoxy-bromobenzene, bromo-3,5-dimethoxybenzene, 3,5-dimethoxy bromobenzene, acmc-1cs59, ksc495e7l
PubChem CID 639187
IUPAC Name 1-bromo-3,5-dimethoxybenzene
SMILES COC1=CC(OC)=CC(Br)=C1

Specifications

Melting Point 62°C to 66°C
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 217.06
Percent Purity 97%
Chemical Name or Material 1-Bromo-3,5-dimethoxybenzene
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only