missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-dihexylbenzene 96.0+%, TCI America™

Catalog No. p-7123771
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48181G 1 g
B48185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48181G Supplier TCI America Supplier No. B48181G
Only null left

Chemical Identifiers

CAS 1238156-36-9
Molecular Formula C18H29Br
Molecular Weight (g/mol) 325.334
InChI Key FVTKYSBFHHVYJG-UHFFFAOYSA-N
PubChem CID 67002258
IUPAC Name 1-bromo-3,5-dihexylbenzene
SMILES CCCCCCC1=CC(=CC(=C1)Br)CCCCCC

Specifications

Color Yellow
Boiling Point 150°C
Quantity 1 g
Formula Weight 325.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3,5-dihexylbenzene