accessibility menu, dialog, popup

UserName

CAS RN 1238156-36-9

H3CBrCH3

CAS RN 1238156-36-9

IUPAC Name: 1-bromo-3,5-dihexylbenzene
Synonyms: 1-bromo-3,5-dihexylbenzene
Molecular Weight (g/mol): 325.33
Molecular Formula: C18H29Br
InChi Key: FVTKYSBFHHVYJG-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC(CCCCCC)=CC(Br)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results