missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3,5-dihexylbenzene 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 150°C |
| Quantity | 5 g |
| Formula Weight | 325.33 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Bromo-3,5-dihexylbenzene |
Safety and Handling
| TSCA | No |