Learn More
1-Bromo-3,5-dichlorobenzene, 97%
CAS: 19752-55-7 | C6H3BrCl2 | 225.894 g/mol
$43.78 - $315.99
Chemical Identifiers
| CAS | 19752-55-7 |
|---|---|
| Molecular Formula | C6H3BrCl2 |
| Molecular Weight (g/mol) | 225.894 |
| MDL Number | MFCD00000584 |
| InChI Key | DZHFFMWJXJBBRG-UHFFFAOYSA-N |
| Synonym | 3,5-dichlorobromobenzene, benzene, 1-bromo-3,5-dichloro, 1-bromo-3,5-dichloro-benzene, 3,5-dichloro bromobenzene, 3,5-dichloro-1-bromobenzene, attercop-chm at113311, maybridge1_000881, pubchem3612, acmc-209f1a |
| PubChem CID | 29766 |
| IUPAC Name | 1-bromo-3,5-dichlorobenzene |
| SMILES | C1=C(C=C(C=C1Cl)Br)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2549206
|
Thermo Scientific Chemicals
B2549206 |
5 g |
Each for $43.78
|
|
|||||
|
AAB2549214
|
Thermo Scientific Chemicals
B2549214 |
25 g |
Each for $115.99
|
|
|||||
|
AAB2549222
|
Thermo Scientific Chemicals
B2549222 |
100 g |
Each for $315.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 19752-55-7 | |
| 225.894 | |
| DZHFFMWJXJBBRG-UHFFFAOYSA-N | |
| 29766 | |
| C1=C(C=C(C=C1Cl)Br)Cl |
| C6H3BrCl2 | |
| MFCD00000584 | |
| 3,5-dichlorobromobenzene, benzene, 1-bromo-3,5-dichloro, 1-bromo-3,5-dichloro-benzene, 3,5-dichloro bromobenzene, 3,5-dichloro-1-bromobenzene, attercop-chm at113311, maybridge1_000881, pubchem3612, acmc-209f1a | |
| 1-bromo-3,5-dichlorobenzene |
Specifications
| 19752-55-7 | |
| 232°C | |
| MFCD00000584 | |
| 1931524 | |
| DZHFFMWJXJBBRG-UHFFFAOYSA-N | |
| 1-bromo-3,5-dichlorobenzene | |
| 29766 | |
| 97% |
| 73°C to 77°C | |
| C6H3BrCl2 | |
| 5 g | |
| 3,5-dichlorobromobenzene, benzene, 1-bromo-3,5-dichloro, 1-bromo-3,5-dichloro-benzene, 3,5-dichloro bromobenzene, 3,5-dichloro-1-bromobenzene, attercop-chm at113311, maybridge1_000881, pubchem3612, acmc-209f1a | |
| C1=C(C=C(C=C1Cl)Br)Cl | |
| 225.894 | |
| 225.91 | |
| 1-Bromo-3,5-dichlorobenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 243-270-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only