missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-dichlorobenzene, 97%

CAS: 19752-55-7 | C6H3BrCl2 | 225.894 g/mol

Supplier:  Thermo Scientific Chemicals B2549214

 View more versions of this product

Catalog No. AAB2549214


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Bromo-3,5-dichlorobenzene
73°C to 77°C
C6H3BrCl2
25 g
3,5-dichlorobromobenzene, benzene, 1-bromo-3,5-dichloro, 1-bromo-3,5-dichloro-benzene, 3,5-dichloro bromobenzene, 3,5-dichloro-1-bromobenzene, attercop-chm at113311, maybridge1_000881, pubchem3612, acmc-209f1a
C1=C(C=C(C=C1Cl)Br)Cl
225.894
225.91
19752-55-7
232°C
MFCD00000584
1931524
DZHFFMWJXJBBRG-UHFFFAOYSA-N
1-bromo-3,5-dichlorobenzene
29766
97%

Chemical Identifiers

19752-55-7
225.894
DZHFFMWJXJBBRG-UHFFFAOYSA-N
29766
C1=C(C=C(C=C1Cl)Br)Cl
C6H3BrCl2
MFCD00000584
3,5-dichlorobromobenzene, benzene, 1-bromo-3,5-dichloro, 1-bromo-3,5-dichloro-benzene, 3,5-dichloro bromobenzene, 3,5-dichloro-1-bromobenzene, attercop-chm at113311, maybridge1_000881, pubchem3612, acmc-209f1a
1-bromo-3,5-dichlorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 243-270-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only