missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-di-tert-butylbenzene, 99%

Catalog No. p-7046507
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1464603 1 g
AAL1464606 5 g
AAL1464614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1464603 Supplier Thermo Scientific Chemicals Supplier No. L1464603
Only null left

CAS: 22385-77-9 | C14H21Br | 269.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22385-77-9
Molecular Formula C14H21Br
Molecular Weight (g/mol) 269.226
MDL Number MFCD00796945
InChI Key BUOWTUULDKULFI-UHFFFAOYSA-N
Synonym 1-bromo-3,5-di-tert-butylbenzene, 3,5-di-tert-butylbromobenzene, 3,5-di-t-butylbromobenzene, 1-bromo-3,5-di-t-butylbenzene, 1-bromo-3,5-ditert-butyl benzene, benzene, 1-bromo-3,5-bis 1,1-dimethylethyl, 1,5-bis tert-butyl-3-bromobenzene, acmc-1cpbo, g00018-watson-int, 3,5-di-t-butyl-bromobenzene
PubChem CID 620136
IUPAC Name 1-bromo-3,5-ditert-butylbenzene
SMILES CC(C)(C)C1=CC(=CC(=C1)Br)C(C)(C)C

Specifications

Melting Point 62°C to 66°C
Quantity 1 g
Beilstein 2091559
Formula Weight 269.23
Percent Purity 99%
Chemical Name or Material 1-Bromo-3,5-di-tert-butylbenzene
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only