missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-bis(trifluoromethyl)benzene 98.0+%, TCI America™

Catalog No. p-7119239
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B175310G 10 g
B175325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B175310G Supplier TCI America Supplier No. B175310G

Chemical Identifiers

CAS 328-70-1
Molecular Formula C8H3BrF6
Molecular Weight (g/mol) 293.006
MDL Number MFCD00000381
InChI Key CSVCVIHEBDJTCJ-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl bromobenzene, 1-bromo-3,5-bis trifluoromethyl benzene, 1,3-bis trifluoromethyl-5-bromobenzene, mbt-br, 3,5-di trifluoromethyl bromobenzene, 3,5-bis trifluoromethyl-1-bromobenzene, 3,5-bis-trifluoromethylbromobenzene, benzene, 1-bromo-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenyl bromide, 1-bromo-3,5-bis-trifluoromethyl-benzene
PubChem CID 67602
IUPAC Name 1-bromo-3,5-bis(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1C(F)(F)F)Br)C(F)(F)F

Specifications

Color Yellow
Boiling Point 151°C
Quantity 10 g
Formula Weight 293.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3,5-bis(trifluoromethyl)benzene