missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-bis(trifluoromethyl)benzene, 98%

Catalog No. p-7082062
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1233709 10 g
AAA1233718 50 g
AAA1233730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1233709 Supplier Thermo Scientific Chemicals Supplier No. A1233709

CAS: 328-70-1 | C8H3BrF6 | 293.006 g/mol

1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 328-70-1
Molecular Formula C8H3BrF6
Molecular Weight (g/mol) 293.006
MDL Number MFCD00000381
InChI Key CSVCVIHEBDJTCJ-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl bromobenzene, 1-bromo-3,5-bis trifluoromethyl benzene, 1,3-bis trifluoromethyl-5-bromobenzene, mbt-br, 3,5-di trifluoromethyl bromobenzene, 3,5-bis trifluoromethyl-1-bromobenzene, 3,5-bis-trifluoromethylbromobenzene, benzene, 1-bromo-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenyl bromide, 1-bromo-3,5-bis-trifluoromethyl-benzene
PubChem CID 67602
IUPAC Name 1-bromo-3,5-bis(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1C(F)(F)F)Br)C(F)(F)F

Specifications

Melting Point -16°C
Density 1.698
Boiling Point 152°C to 155°C
Flash Point >110°C (230°F)
Refractive Index 1.427
Quantity 10 g
Beilstein 2123669
Solubility Information Immiscible with water.
Formula Weight 293.01
Percent Purity 98%
Chemical Name or Material 1-Bromo-3,5-bis (trifluoromethyl)benzene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-334-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only