missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-naphthol, 97%

Catalog No. AAA1869222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1869222 100 g
AAA1869214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1869222 Supplier Thermo Scientific Chemicals Supplier No. A1869222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 573-97-7 | C10H7BrO | 223.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 573-97-7
Molecular Formula C10H7BrO
Molecular Weight (g/mol) 223.069
MDL Number MFCD00003869
InChI Key FQJZPYXGPYJJIH-UHFFFAOYSA-N
Synonym 1-bromo-2-naphthol, 1-bromo-2-naphthalenol, wormin, 2-naphthalenol, 1-bromo, disthemin, 1-bromo-2-hydroxynaphthalene, 1-brom-2-naphthol, unii-vke7l55pp9, 1-brom-2-naphthol german, 2-naphthol, 1-bromo
PubChem CID 11316
IUPAC Name 1-bromonaphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2Br)O

Specifications

Melting Point 76°C to 81°C
Quantity 100 g
Beilstein 2044001
Formula Weight 223.08
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-naphthol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-363-1
RTECSNumber QL3361000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only