accessibility menu, dialog, popup

UserName

HOBr

CAS RN 573-97-7

IUPAC Name: 1-bromonaphthalen-2-ol
Synonyms: 1-bromo-2-hydroxynaphthalene
Molecular Weight (g/mol): 223.07
Molecular Formula: C10H7BrO
InChi Key: FQJZPYXGPYJJIH-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC=CC2=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)