missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methylpropane 97.0+%, TCI America™

Catalog No. B061625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B061625G 25 g
B0616500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B061625G Supplier TCI America Supplier No. B061625G

Chemical Identifiers

CAS 78-77-3
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000217
InChI Key HLVFKOKELQSXIQ-UHFFFAOYSA-N
Synonym isobutyl bromide, iso-butyl bromide, propane, 1-bromo-2-methyl, bromoisobutane, i-butyl bromide, 1-bromo-2-methyl-propane, unii-5oec0bw987, ccris 349, 1-bromo-2-methyl propane, bromoacetaldehydedimethylacetal
PubChem CID 6555
IUPAC Name 1-bromo-2-methylpropane
SMILES CC(C)CBr

Specifications

Color Yellow
Boiling Point 92°C
Quantity 25 g
UN Number 2342
Formula Weight 137.02
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-methylpropane
EINECSNumber (2)-0074
RTECSNumber TX4140000
TSCA Yes