missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methylpropane, 98+%

Catalog No. p-4510470
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168831000 100 mL Glass bottle
AC168835000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC168831000 Supplier Thermo Scientific Chemicals Supplier No. 168831000
Only null left

CAS: 78-77-3 | C4H9Br | 137.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-77-3
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
InChI Key HLVFKOKELQSXIQ-UHFFFAOYSA-N
Synonym isobutyl bromide, iso-butyl bromide, propane, 1-bromo-2-methyl, bromoisobutane, i-butyl bromide, 1-bromo-2-methyl-propane, unii-5oec0bw987, ccris 349, 1-bromo-2-methyl propane, bromoacetaldehydedimethylacetal
PubChem CID 6555
IUPAC Name 1-bromo-2-methylpropane
SMILES CC(C)CBr

Specifications

Melting Point -119.0°C
Color Colorless
Density 1.2600g/mL
Boiling Point 90.0°C to 92.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4346 to 1.4366
Linear Formula (CH3)2CHCH2Br
Quantity 100 mL
Beilstein 01,126
Merck Index 15,518
Specific Gravity 1.26
Solubility Information Solubility in water: 0.51g/L (18°C). Other solubilities: soluble in most common organic solvents
Formula Weight 137.02
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-methylpropane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protect
EINECSNumber 201-141-2

RUO – Research Use Only