missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-iodobenzene (stabilized with Copper chip) 98.0+%, TCI America™
Supplier: TCI America B1126250G
Specifications
| 1-Bromo-2-iodobenzene (stabilized with Copper chip) | |
| 120°C | |
| MFCD00001030 | |
| 2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene | |
| C1=CC=C(C(=C1)Br)I | |
| 282.906 | |
| 282.91 | |
| Liquid |
| 583-55-1 | |
| C6H4BrI | |
| 250 g | |
| OIRHKGBNGGSCGS-UHFFFAOYSA-N | |
| 1-bromo-2-iodobenzene | |
| 11415 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 583-55-1 | |
| 282.906 | |
| OIRHKGBNGGSCGS-UHFFFAOYSA-N | |
| 11415 | |
| C1=CC=C(C(=C1)Br)I |
| C6H4BrI | |
| MFCD00001030 | |
| 2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene | |
| 1-bromo-2-iodobenzene |
Safety and Handling
TSCA : Yes