accessibility menu, dialog, popup

UserName

BrI

CAS RN 583-55-1

IUPAC Name: 1-bromo-2-iodobenzene
Synonyms: benzene, 1-bromo-2-iodo-
Molecular Weight (g/mol): 282.91
Molecular Formula: C6H4BrI
InChi Key: OIRHKGBNGGSCGS-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1I
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (5)