Learn More
1-Bromo-2-iodobenzene, 98+%, stab. with copper
CAS: 583-55-1 | C6H4BrI | 282.906 g/mol
Supplier: Thermo Scientific Chemicals A1702014
Description
2-Bromoiodobenzene is used in the synthesis of diarylamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromoiodobenzene is used in the synthesis of diarylamines.
Solubility
Not miscible or difficult to mix in water.
Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.
Specifications
| 1-Bromo-2-iodobenzene | |
| 583-55-1 | |
| 2.256 | |
| >110°C (230°F) | |
| 1.6645 | |
| 25 g | |
| Light sensitive | |
| Not miscible or difficult to mix in water. | |
| C1=CC=C(C(=C1)Br)I | |
| 282.906 | |
| 282.91 |
| stabilized with copper | |
| 9°C to 10°C | |
| 257°C | |
| C6H4BrI | |
| MFCD00001030 | |
| 1855309 | |
| 2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene | |
| OIRHKGBNGGSCGS-UHFFFAOYSA-N | |
| 1-bromo-2-iodobenzene | |
| 11415 | |
| ≥98% |
Chemical Identifiers
| 583-55-1 | |
| 282.906 | |
| OIRHKGBNGGSCGS-UHFFFAOYSA-N | |
| 11415 | |
| C1=CC=C(C(=C1)Br)I |
| C6H4BrI | |
| MFCD00001030 | |
| 2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene | |
| 1-bromo-2-iodobenzene |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-508-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only