missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-iodobenzene, 98+%, stab. with copper

Catalog No. AAA1702022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1702022 100 g
AAA1702006 5 g
AAA1702014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1702022 Supplier Thermo Scientific Chemicals Supplier No. A1702022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 583-55-1 | C6H4BrI | 282.906 g/mol

2-Bromoiodobenzene is used in the synthesis of diarylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoiodobenzene is used in the synthesis of diarylamines.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.

Chemical Identifiers

CAS 583-55-1
Molecular Formula C6H4BrI
Molecular Weight (g/mol) 282.906
MDL Number MFCD00001030
InChI Key OIRHKGBNGGSCGS-UHFFFAOYSA-N
Synonym 2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene
PubChem CID 11415
IUPAC Name 1-bromo-2-iodobenzene
SMILES C1=CC=C(C(=C1)Br)I

Specifications

Melting Point 9°C to 10°C
Density 2.256
Boiling Point 257°C
Flash Point >110°C (230°F)
Refractive Index 1.6645
Quantity 100 g
Beilstein 1855309
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 282.91
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-2-iodobenzene, stabilized with copper
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-508-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only