missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-fluoro-4-iodobenzene 97.0+%, TCI America™
$78.93 - $232.95
Chemical Identifiers
| CAS | 136434-77-0 |
|---|---|
| Molecular Formula | C6H3BrFI |
| Molecular Weight (g/mol) | 300.90 |
| MDL Number | MFCD00042577 |
| InChI Key | OCODJNASCDFXSR-UHFFFAOYSA-N |
| Synonym | 4-bromo-3-fluoroiodobenzene, 3-fluoro-4-bromo iodobenzene, 1-bromo-2-fluoro-4-iodo-benzene, benzene, 1-bromo-2-fluoro-4-iodo, 3-fluoro-4-bromo-iodobenzene, 1-bromo-2-fluoro-4-i, pubchem3231, acmc-209c5x, 3-fluoro-4-bromoiodobenzene, ksc490o1h |
| PubChem CID | 2725006 |
| IUPAC Name | 1-bromo-2-fluoro-4-iodobenzene |
| SMILES | FC1=C(Br)C=CC(I)=C1 |
Chemical Identifiers
| 136434-77-0 | |
| 300.90 | |
| OCODJNASCDFXSR-UHFFFAOYSA-N | |
| 2725006 | |
| FC1=C(Br)C=CC(I)=C1 |
| C6H3BrFI | |
| MFCD00042577 | |
| 4-bromo-3-fluoroiodobenzene, 3-fluoro-4-bromo iodobenzene, 1-bromo-2-fluoro-4-iodo-benzene, benzene, 1-bromo-2-fluoro-4-iodo, 3-fluoro-4-bromo-iodobenzene, 1-bromo-2-fluoro-4-i, pubchem3231, acmc-209c5x, 3-fluoro-4-bromoiodobenzene, ksc490o1h | |
| 1-bromo-2-fluoro-4-iodobenzene |
Specifications
| 136434-77-0 | |
| 112°C | |
| MFCD00042577 | |
| 4-bromo-3-fluoroiodobenzene, 3-fluoro-4-bromo iodobenzene, 1-bromo-2-fluoro-4-iodo-benzene, benzene, 1-bromo-2-fluoro-4-iodo, 3-fluoro-4-bromo-iodobenzene, 1-bromo-2-fluoro-4-i, pubchem3231, acmc-209c5x, 3-fluoro-4-bromoiodobenzene, ksc490o1h | |
| FC1=C(Br)C=CC(I)=C1 | |
| 300.90 | |
| 300.90 | |
| Crystalline Powder |
| 39°C | |
| C6H3BrFI | |
| 5 g | |
| OCODJNASCDFXSR-UHFFFAOYSA-N | |
| 1-bromo-2-fluoro-4-iodobenzene | |
| 2725006 | |
| ≥97.0% (GC) | |
| 1-Bromo-2-fluoro-4-iodobenzene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator