accessibility menu, dialog, popup

UserName

IFBr

CAS RN 136434-77-0

IUPAC Name: 1-bromo-2-fluoro-4-iodobenzene
Synonyms: 1-bromo-2-fluoro-4-iodobenzene
Molecular Weight (g/mol): 300.9
Molecular Formula: C6H3BrFI
InChi Key: OCODJNASCDFXSR-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=CC(I)=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)