missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-fluoro-4-iodobenzene 97.0+%, TCI America™

Catalog No. B30175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30175G 5 g
B301725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30175G Supplier TCI America Supplier No. B30175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 136434-77-0
Molecular Formula C6H3BrFI
Molecular Weight (g/mol) 300.90
MDL Number MFCD00042577
InChI Key OCODJNASCDFXSR-UHFFFAOYSA-N
Synonym 4-bromo-3-fluoroiodobenzene, 3-fluoro-4-bromo iodobenzene, 1-bromo-2-fluoro-4-iodo-benzene, benzene, 1-bromo-2-fluoro-4-iodo, 3-fluoro-4-bromo-iodobenzene, 1-bromo-2-fluoro-4-i, pubchem3231, acmc-209c5x, 3-fluoro-4-bromoiodobenzene, ksc490o1h
PubChem CID 2725006
IUPAC Name 1-bromo-2-fluoro-4-iodobenzene
SMILES FC1=C(Br)C=CC(I)=C1

Specifications

Melting Point 39°C
Boiling Point 112°C
Quantity 5 g
Formula Weight 300.90
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2-fluoro-4-iodobenzene
TSCA No
Recommended Storage Refrigerator