missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-ethynylbenzene 98.0+%, TCI America™

Catalog No. B46081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46081G 1 g
B46085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46081G Supplier TCI America Supplier No. B46081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 766-46-1
Molecular Formula C8H5Br
Molecular Weight (g/mol) 181.032
MDL Number MFCD01863358
InChI Key RVDOYUFNRDGYGU-UHFFFAOYSA-N
Synonym 2-Bromophenylacetylene
PubChem CID 602067
IUPAC Name 1-bromo-2-ethynylbenzene
SMILES C#CC1=CC=CC=C1Br

Specifications

Boiling Point 94°C
Quantity 1 g
Formula Weight 181.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-ethynylbenzene