CAS RN 766-46-1
CAS RN 766-46-1
IUPAC Name:
1-bromo-2-ethynylbenzene
Synonyms:
1-bromo-2-ethynylbenzene
Molecular Weight (g/mol):
181.03
Molecular Formula:
C8H5Br
InChi Key:
RVDOYUFNRDGYGU-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC=C1C#C
1
–
1
of
1
results
1
1-Bromo-2-ethynylbenzene 98.0+%, TCI America™
CAS: 766-46-1 Molecular Formula: C8H5Br Molecular Weight (g/mol): 181.032 MDL Number: MFCD01863358 InChI Key: RVDOYUFNRDGYGU-UHFFFAOYSA-N Synonym: 2-Bromophenylacetylene PubChem CID: 602067 IUPAC Name: 1-bromo-2-ethynylbenzene SMILES: C#CC1=CC=CC=C1Br