missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-ethylhexane 97.0+%, TCI America™

Catalog No. p-7118553
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B059625ML 25 mL
B0596100ML 100 mL
B0596500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B059625ML Supplier TCI America Supplier No. B059625ML

Chemical Identifiers

CAS 18908-66-2
Molecular Formula C8H17Br
Molecular Weight (g/mol) 193.128
MDL Number MFCD00000220
InChI Key NZWIYPLSXWYKLH-UHFFFAOYSA-N
Synonym 2-ethylhexyl bromide, 3-bromomethyl heptane, heptane, 3-bromomethyl, 1-bromo-2-ethylhexane, 1-bromo iso octane, 2-ethyl-bromohexane, 3-bromomethyl-heptane, 1-bromo-2 ethylhexane, 2-ethylhexylhydrobromide
PubChem CID 86804
IUPAC Name 3-(bromomethyl)heptane
SMILES CCCCC(CC)CBr

Specifications

Color Yellow
Boiling Point 156°C
Quantity 25 mL
Formula Weight 193.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-ethylhexane
EINECSNumber (2)-0067
TSCA Yes