missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-ethylhexane 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 156°C |
| Quantity | 100 mL |
| Formula Weight | 193.13 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Bromo-2-ethylhexane |
Safety and Handling
| EINECSNumber | (2)-0067 |
| TSCA | Yes |