missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-chloro-4-nitrobenzene 97.0+%, TCI America™
Supplier: TCI America B534825G
Specifications
| 1-Bromo-2-chloro-4-nitrobenzene | |
| 61°C | |
| C6H3BrClNO2 | |
| 25 g | |
| YKSXEJZFIQAUHJ-UHFFFAOYSA-N | |
| 1-bromo-2-chloro-4-nitrobenzene | |
| 34667 | |
| ≥97.0% (GC) |
| 29682-39-1 | |
| Yellow | |
| MFCD00051514 | |
| 4-bromo-3-chloronitrobenzene, benzene,1-bromo-2-chloro-4-nitro, pubchem22601, acmc-209vlc, 2-chloro-4-nitro-bromobenzene, 4-bromo-3-chloro-nitrobenzene, bromo-2-chloro-4-nitrobenzene, 1-bromanyl-2-chloranyl-4-nitro-benzene | |
| C1=CC(=C(C=C1[N+](=O)[O-])Cl)Br | |
| 236.449 | |
| 236.45 | |
| Crystalline Powder |
Chemical Identifiers
| 29682-39-1 | |
| 236.449 | |
| YKSXEJZFIQAUHJ-UHFFFAOYSA-N | |
| 34667 | |
| C1=CC(=C(C=C1[N+](=O)[O-])Cl)Br |
| C6H3BrClNO2 | |
| MFCD00051514 | |
| 4-bromo-3-chloronitrobenzene, benzene,1-bromo-2-chloro-4-nitro, pubchem22601, acmc-209vlc, 2-chloro-4-nitro-bromobenzene, 4-bromo-3-chloro-nitrobenzene, bromo-2-chloro-4-nitrobenzene, 1-bromanyl-2-chloranyl-4-nitro-benzene | |
| 1-bromo-2-chloro-4-nitrobenzene |
Safety and Handling
TSCA : No