missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloro-4-nitrobenzene 97.0+%, TCI America™

Catalog No. p-7124050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53485G 5 g
B534825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53485G Supplier TCI America Supplier No. B53485G
Only null left

Chemical Identifiers

CAS 29682-39-1
Molecular Formula C6H3BrClNO2
Molecular Weight (g/mol) 236.449
MDL Number MFCD00051514
InChI Key YKSXEJZFIQAUHJ-UHFFFAOYSA-N
Synonym 4-bromo-3-chloronitrobenzene, benzene,1-bromo-2-chloro-4-nitro, pubchem22601, acmc-209vlc, 2-chloro-4-nitro-bromobenzene, 4-bromo-3-chloro-nitrobenzene, bromo-2-chloro-4-nitrobenzene, 1-bromanyl-2-chloranyl-4-nitro-benzene
PubChem CID 34667
IUPAC Name 1-bromo-2-chloro-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)Br

Specifications

Melting Point 61°C
Color Yellow
Quantity 5 g
Formula Weight 236.45
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2-chloro-4-nitrobenzene