missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloro-4-fluorobenzene 98.0+%, TCI America™

Catalog No. p-7122893
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33495G 5 g
B334925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33495G Supplier TCI America Supplier No. B33495G

Chemical Identifiers

CAS 110407-59-5
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD00142587
InChI Key LEFQPBAWVJEIJS-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobromobenzene, 4-bromo-3-chloro-1-fluorobenzene, 1-bromo-2-chloro-4-fluoro-benzene, 4-bromo-3-chlorofluorobenzene, 2-bromo-5-fluorochlorobenzene, benzene, 1-bromo-2-chloro-4-fluoro, pubchem3755, acmc-1bnv0, 2-chloro-4-fluorobromobezene, ksc494m7f
PubChem CID 2733406
IUPAC Name 1-bromo-2-chloro-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)Cl)Br

Specifications

Color Yellow
Quantity 5 g
Formula Weight 209.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-chloro-4-fluorobenzene