accessibility menu, dialog, popup

UserName

FClBr

CAS RN 110407-59-5

IUPAC Name: 1-bromo-2-chloro-4-fluorobenzene
Synonyms: 1-bromo-2-chloro-4-fluorobenzene
Molecular Weight (g/mol): 209.44
Molecular Formula: C6H3BrClF
InChi Key: LEFQPBAWVJEIJS-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=C(Br)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)