missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,6-dichlorobenzene 98.0+%, TCI America™

Catalog No. B09825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09825G 5 g
B098225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09825G Supplier TCI America Supplier No. B09825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19393-92-1
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.894
MDL Number MFCD00000574
InChI Key UWOIDOQAQPUVOH-UHFFFAOYSA-N
Synonym 1-bromo-2,6-dichlorobenzene, 2,6-dichlorobromobenzene, 1,3-dichloro-2-bromobenzene, benzene, 2-bromo-1,3-dichloro, 2,6-dichloro-1-bromobenzene, benzene, 1-bromo-2,6-dichloro, pubchem3610, acmc-209exg, 2,6-dichlorobromo benzene, 2,6-dichlorophenyl bromide
PubChem CID 29568
IUPAC Name 2-bromo-1,3-dichlorobenzene
SMILES C1=CC(=C(C(=C1)Cl)Br)Cl

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 242°C
Quantity 5 g
Formula Weight 225.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2,6-dichlorobenzene