accessibility menu, dialog, popup

UserName

CAS RN 19393-92-1

IUPAC Name: 2-bromo-1,3-dichlorobenzene
Synonyms: 1-bromo-2,6-dichlorobenzene
Molecular Weight (g/mol): 225.89
Molecular Formula: C6H3BrCl2
InChi Key: UWOIDOQAQPUVOH-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1Br
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)