missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,5-dimethoxybenzene 97.0+%, TCI America™

Catalog No. p-7119396
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19795G 5 g
B197925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19795G Supplier TCI America Supplier No. B19795G

Chemical Identifiers

CAS 25245-34-5
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD00008355
InChI Key DWCGNRKFLRLWCJ-UHFFFAOYSA-N
PubChem CID 91353
IUPAC Name 2-bromo-1,4-dimethoxybenzene
SMILES COC1=CC=C(OC)C(Br)=C1

Specifications

Color Yellow
Boiling Point 237°C
Quantity 5 g
Synonym 1-bromo-2,5-dimethoxybenzene, benzene, 2-bromo-1,4-dimethoxy, 2,5-dimethoxybromobenzene, pubchem19785, 2-bromo-4-methoxyanisole, acmc-1cby3, ksc490m9p, 1,4-dimethoxy-2-bromobenzene, 1-bromo-2, 5-dimethoxybenzene, dwcgnrkflrlwcj-uhfffaoysa
Formula Weight 217.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2,5-dimethoxybenzene