accessibility menu, dialog, popup

UserName

H3COOCH3Br

CAS RN 25245-34-5

IUPAC Name: 2-bromo-1,4-dimethoxybenzene
Synonyms: 2-bromo-1,4-dimethoxybenzene
Molecular Weight (g/mol): 217.06
Molecular Formula: C8H9BrO2
InChi Key: DWCGNRKFLRLWCJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(Br)=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)