missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,5-difluorobenzene 98.0+%, TCI America™

Catalog No. p-7118892
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11665G 5 g
B116625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11665G Supplier TCI America Supplier No. B11665G

Chemical Identifiers

CAS 399-94-0
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.991
MDL Number MFCD00000345
InChI Key XCRCSPKQEDMVBO-UHFFFAOYSA-N
Synonym 1-bromo-2,5-difluorobenzene, 2,5-difluorobromobenzene, benzene, 2-bromo-1,4-difluoro, 1,4-difluoro-2-bromobenzene, 2-bromo-1,4-difluoro-benzene, fr df be, pubchem2302, acmc-1ad5i, 1-bromo-2,5 difluorobenzene, 2,5-difluoro-1-bromobenzene
PubChem CID 67862
IUPAC Name 2-bromo-1,4-difluorobenzene
SMILES C1=CC(=C(C=C1F)Br)F

Specifications

Color Yellow
Boiling Point 58°C
Quantity 5 g
Formula Weight 192.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2,5-difluorobenzene