missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2,5-bis(trifluoromethyl)benzene 87.0+%, TCI America™
Supplier: TCI America B254525G
Specifications
| 1-Bromo-2,5-bis(trifluoromethyl)benzene | |
| 5°C | |
| C8H3BrF6 | |
| 25 g | |
| GFQNSGHVOFVTLC-UHFFFAOYSA-N | |
| 2-bromo-1,4-bis(trifluoromethyl)benzene | |
| 2734763 | |
| ≥87.0% (GC) |
| 7617-93-8 | |
| 158°C | |
| MFCD03094336 | |
| 2,5-bis trifluoromethyl bromobenzene, 2-bromo-1,4-bis trifluoromethyl benzene, 1-bromo-2,5-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-bromobenzene, benzene, 2-bromo-1,4-bis trifluoromethyl, pubchem4264, acmc-1bisb, ksc495k7j, 2,5-bis-trifluoromethylbromobenzene | |
| C1=CC(=C(C=C1C(F)(F)F)Br)C(F)(F)F | |
| 293.006 | |
| 293.01 | |
| Liquid |
Chemical Identifiers
| 7617-93-8 | |
| 293.006 | |
| GFQNSGHVOFVTLC-UHFFFAOYSA-N | |
| 2734763 | |
| C1=CC(=C(C=C1C(F)(F)F)Br)C(F)(F)F |
| C8H3BrF6 | |
| MFCD03094336 | |
| 2,5-bis trifluoromethyl bromobenzene, 2-bromo-1,4-bis trifluoromethyl benzene, 1-bromo-2,5-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-bromobenzene, benzene, 2-bromo-1,4-bis trifluoromethyl, pubchem4264, acmc-1bisb, ksc495k7j, 2,5-bis-trifluoromethylbromobenzene | |
| 2-bromo-1,4-bis(trifluoromethyl)benzene |
Safety and Handling
TSCA : No