CAS RN 7617-93-8
IUPAC Name:
2-bromo-1,4-bis(trifluoromethyl)benzene
Synonyms:
2-bromo-1,4-bis(trifluoromethyl)benzene
Molecular Weight (g/mol):
293.01
Molecular Formula:
C8H3BrF6
InChi Key:
GFQNSGHVOFVTLC-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(C(Br)=C1)C(F)(F)F
1
–
1
of
1
results
1
1-Bromo-2,5-bis(trifluoromethyl)benzene 87.0+%, TCI America™
CAS: 7617-93-8 Molecular Formula: C8H3BrF6 Molecular Weight (g/mol): 293.006 MDL Number: MFCD03094336 InChI Key: GFQNSGHVOFVTLC-UHFFFAOYSA-N Synonym: 2,5-bis trifluoromethyl bromobenzene,2-bromo-1,4-bis trifluoromethyl benzene,1-bromo-2,5-bis trifluoromethyl benzene,1,4-bis trifluoromethyl-2-bromobenzene,benzene, 2-bromo-1,4-bis trifluoromethyl,pubchem4264,acmc-1bisb,ksc495k7j,2,5-bis-trifluoromethylbromobenzene PubChem CID: 2734763 IUPAC Name: 2-bromo-1,4-bis(trifluoromethyl)benzene SMILES: C1=CC(=C(C=C1C(F)(F)F)Br)C(F)(F)F