missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-dimethoxybenzene, 98%

Catalog No. AAA1443914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1443914 25 g
AAA1443922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1443914 Supplier Thermo Scientific Chemicals Supplier No. A1443914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17715-69-4 | C8H9BrO2 | 217.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17715-69-4
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD00009844
InChI Key NIUZVSQOXJIHBL-UHFFFAOYSA-N
Synonym 2,4-dimethoxybromobenzene, 1,3-dimethoxy-4-bromobenzene, 4-bromo-3-methoxyanisole, benzene, 1-bromo-2,4-dimethoxy, 4-bromoresorcinol dimethyl ether, 4-bromoresocinol dimethyl ether, 4-bromo-1,3-dimethoxybenzene, 1-brom-2,4-dimethoxybenzol, pubchem3036, acmc-1c5dw
PubChem CID 87266
IUPAC Name 1-bromo-2,4-dimethoxybenzene
SMILES COC1=CC=C(Br)C(OC)=C1

Specifications

Melting Point 24°C to 26°C
Density 1.496
Boiling Point 153°C to 155°C (18 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.572
Quantity 25 g
Beilstein 1867593
Formula Weight 217.07
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,4-dimethoxybenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-717-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only