accessibility menu, dialog, popup

UserName

H3COBrOH3C

CAS RN 17715-69-4

IUPAC Name: 1-bromo-2,4-dimethoxybenzene
Synonyms: 1-bromo-2,4-dimethoxybenzene
Molecular Weight (g/mol): 217.06
Molecular Formula: C8H9BrO2
InChi Key: NIUZVSQOXJIHBL-UHFFFAOYSA-N
SMILES: COC1=CC=C(Br)C(OC)=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)