missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2,4-difluoro-5-nitrobenzene 98.0+%, TCI America™
$38.51 - $100.11
Chemical Identifiers
| CAS | 345-24-4 |
|---|---|
| Molecular Formula | C6H2BrF2NO2 |
| Molecular Weight (g/mol) | 237.99 |
| MDL Number | MFCD00129166 |
| InChI Key | OOUUWURPSUSDTD-UHFFFAOYSA-N |
| Synonym | 5-Bromo-2,4-difluoronitrobenzene |
| PubChem CID | 223078 |
| IUPAC Name | 1-bromo-2,4-difluoro-5-nitrobenzene |
| SMILES | [O-][N+](=O)C1=CC(Br)=C(F)C=C1F |
Chemical Identifiers
| 345-24-4 | |
| 237.99 | |
| OOUUWURPSUSDTD-UHFFFAOYSA-N | |
| 223078 | |
| [O-][N+](=O)C1=CC(Br)=C(F)C=C1F |
| C6H2BrF2NO2 | |
| MFCD00129166 | |
| 5-Bromo-2,4-difluoronitrobenzene | |
| 1-bromo-2,4-difluoro-5-nitrobenzene |
Specifications
| 345-24-4 | |
| Yellow | |
| C6H2BrF2NO2 | |
| 1 g | |
| OOUUWURPSUSDTD-UHFFFAOYSA-N | |
| 1-bromo-2,4-difluoro-5-nitrobenzene | |
| 223078 | |
| ≥98.0% (GC) | |
| 1-Bromo-2,4-difluoro-5-nitrobenzene |
| 19°C | |
| 97°C | |
| MFCD00129166 | |
| 5-Bromo-2,4-difluoronitrobenzene | |
| [O-][N+](=O)C1=CC(Br)=C(F)C=C1F | |
| 237.99 | |
| 237.99 | |
| Liquid |
Safety and Handling
TSCA : No